Wall Mounted Draining Racks, Bürkle
À propos de cet article
This flexible PVC system has stable drainage pegs, available in three lengths (60, 100 and 150 mm) with Ø 6, 10 and 12 mm, that can be repositioned at any time. The draining racks can be adapted to any apparatus which is to be dried.
- Will not shatter or break
- Resistant to most chemicals
- Removable drainage channel with connection for a draining tube hose (for drainage of dripping water into laboratory basin)
Ordering information: Rack supplied complete with removable dust- and scratch-proof protective film, and rods, screws and dowels for wall mounting.
6 Options Available Below
Product Details & Documents
This flexible PVC system has stable drainage pegs, available in three lengths (60, 100 and 150 mm) with Ø 6, 10 and 12 mm, that can be repositioned at any time. The draining racks can be adapted to any apparatus which is to be dried.
- Will not shatter or break
- Resistant to most chemicals
- Removable drainage channel with connection for a draining tube hose (for drainage of dripping water into laboratory basin)
Ordering information: Rack supplied complete with removable dust- and scratch-proof protective film, and rods, screws and dowels for wall mounting.
Product Family Options
Draining Racks with Drainage Channels
Informations sur le produit
- DésignationHauteurLargeurAvailabilityPrice
- Supplier Number: 9640-2040Spécifications:
More
Specifications
Cat. No.10217-314 - Supplier Number: 9640-2050Spécifications:
More
Specifications
Cat. No.10217-316 - Supplier Number: 9640-2060Spécifications:
More
Specifications
Cat. No.10217-318
Draining Racks without Drainage Channels
Informations sur le produit
- DésignationHauteurLargeurAvailabilityPrice
- Supplier Number: 9640-1040Spécifications:
More
Specifications
Cat. No.10217-308 - Supplier Number: 9640-1050Spécifications:
More
Specifications
Cat. No.10217-310 - Supplier Number: 9640-1060Spécifications:
More
Specifications
Cat. No.10217-312
Accessories
Informations sur le produit
- Catalog #LengthAvailabilityPrice
- Rods, PP, Ø39/100"
Informations sur le produit
- Catalog #LengthAvailabilityPrice
- Rods, PP, Ø 47/100
Informations sur le produit
- Catalog #LengthAvailabilityPrice
- Rods, PP, Ø 23/100