Sub Surface Sampler, Grab Sampler® I
Sub Surface Grab Sampler®, 1000 ml
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 990250
Jump to:
Product Details & Documents
Ideal for sampling from spillways, docks and other sub surfaces.
- Lightweight construction
- Eliminates surface contamination
- No need to physically enter the sample area
- Helps meet EPA requirements for water and wastewater sampling
The basic unit features a 183 cm aluminum body with clamp for securing a 1000 ml sample bottle. An attached extension allows the bottle to be opened and closed under water for discrete sampling.
Documents
Spécifications
- Capacité:1000 ml
- Désignation:Sub Surface Grab Sampler®
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- 1000mL Bottle with Cap size of 38-430