Microtest Manifolds, DWK Life Sciences
These autoclavable stainless steel microtest manifolds are ideal for use with 24 or 96 well microplates
It tightly fit with both Luer and Luer Lock syringe nozzles. The manifolda available in both a straight "T" style and an angular style with an offset hub.
Frequently Bought Together