Chromatography Gravity Columns with Standard Taper Joint and Fritted Disk, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Gravity chromatography column manufactured from medium wall tubing and in-line reservoir for containing liquid buffer.
Standard Taper joint size 24/40. A coarse porosity glass frit (40 to 60 µm pore size) is a support for column packing material. Custom sizes and plastic safety coating are available upon request.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: PTFE stopcock needs to be purchased separately.
Frequently Bought Together