Precision Zip-Clean Pouches™, PPC Flexible Packaging
Tough, ultraclean, high clarity film that is ideal for all multi-use, reclosable cleanroom and laboratory applications.
- Easy open and easy zip closure
- Cleanroom certified
- Barrier properties
Sealed in a double-lined carton.
Frequently Bought Together