Chromatography Glass Columns with Reservoir, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Gravity chromatography column manufactured from medium wall tubing with PTFE stopcock and in-line reservoir for containing liquid buffer.
- Standard taper joint size 24/40
Glass wool (not supplied) may be used as a support for the packing material. Custom sizes and plastic safety coating are available upon request.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together