Snap Caps, Wheaton
Snap Caps, Wheaton
Snap Caps, Wheaton
Certificates
Snap Caps, Wheaton
Snap Caps, Wheaton
4 Options Available Below
Product Details & Documents
Eliminate the need for crimping and decapping tools.
  • Manufactured from polyethylene
Fits the WHEATON E-Z vials® with snap ring top.
Product Family Options
With PTFE / Red Rubber Septa
Product Information
  • Désignation
    Taille
    Septum
    Availability
    Price
  • Natural cap
    11 mm
    PTFE/Red Rubber
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $871.15
    $871.15
    New Customer Promotional Price
    Case of 1,000
       $871.15
    Supplier Number: 242786
With PTFE / Silicone Septa
Product Information
  • Désignation
    Taille
    Septum
    Availability
    Price
  • Natural cap with cross slit
    11 mm
    PTFE/Silicone
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $1,786.00
    $1,786.00
    New Customer Promotional Price
    Case of 1,000
       $1,786.00
    Supplier Number: 242775
  • Natural cap
    11 mm
    PTFE/Silicone
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $1,263.50
    $1,263.50
    New Customer Promotional Price
    Case of 1,000
       $1,263.50
    Supplier Number: 242776
With Red PTFE / Silicone / Red PTFE Septa
Product Information
  • Désignation
    Taille
    Septum
    Availability
    Price
  • Natural cap
    11 mm
    Red PTFE/Silicone
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $1,529.50
    $1,529.50
    New Customer Promotional Price
    Case of 1,000
       $1,529.50
    Supplier Number: 242772