Bessman Tissue Pulverizer, Spectrum® Laboratories
The stainless steel Bessman Tissue Pulverizer consists of a 2 component mortar with handles and a pestle, specifically designed for pulverizing 10 - 250 mg of tissue.
- 2 sizes 10 - 50 mg and 25 - 250 mg available
- Includes 10 test tubes
After chilling the mortar and pestle in liquid nitrogen, the included 28 cm lead hammer is used on the mortar to completely pulverize the inserted sample. The sample can then easily be transferred without touching or thawing by removing the mortar and inverting the pestle assembly over a test tube.
Frequently Bought Together