Simultaneous Test Sets, Drager Safety
 :   Draeger
Simultaneous Test Sets, Drager Safety
 :  Draeger
Simultaneous Test Sets, Drager Safety
Simultaneous Test Sets, Drager Safety
Risks and potential dangers at a glance: The SimultanTest Sets allow the measurement of five different gases at the same time.
In the SimultanTest Set, five Dräger-Tubes are arranged next to each other in a stable rubber sleeve. The air to be monitored is simultaneously drawn in through the tubes by a Dräger-Tube pump. Quick determinations are made about the presence of dangerous substances at TLV levels.
The device delivers fast and reliable results for different combinations of organic and inorganic gases.
Product Family Options
  • Désignation
    Response Time
  • Simultaneous test set I, for inorganic fumes (acid gas, hydrocyanic acid, carbon monoxide, basic gas, nitrous gas)
    40 s
    Pack of 10
    Pack of 10 - $658.92
    $658.92
    Pack of 10
       $658.92
     : 8101735
  • Simultaneous test set III, for organic vapors (ketone, aromates, alcohol, aliphatic hydrocarbons, chlorinated hydrocarbons)
    120 s
    Pack of 10
    Pack of 10 - $658.92
    $658.92
    Pack of 10
       $658.92
     : 8101770
  • Simultaneous test set II, for inorganic fumes (sulfur dioxide, chlorine, hydrogen sulfide, hydrogen phosphide, carbonyl chloride)
    40 s
    Each
    Each - $164.37
    $164.37
    Each
       $164.37
     : 8101736
Accessories
  •  
    Description
  • CA13450-858
    Extension Hose
    Each - $1,769.37
  •  
    Description
  • CA34330-044
    Adapter for Simultaneous test sets, for connection to a bellows pump
    Each - $595.27