Spectra/Por®, Dialysis Tubing, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Certificates
About this item
The dialysis tubing is sealed at one end with a closure and open at the other.
Supplied wet in 0.1% sodium azide. Should be rinsed prior to use.
Length: 600 mm
A funnel is attached to the top of the sack for easily filling.
Frequently Bought Together