I.V. Training Hand | Avantor
I.V. Training Hand
I.V. Training Hand
Certificates
I.V. Training Hand
I.V. Training Hand
3 Options Available Below
Product Details & Documents
Basic Hand Injection Practice
  • Basic I.V. training introduction
  • Supple skin allows for vein palpation
This training aid was designed for practice in giving IVs. It permits detailed exercises in venipuncture so that the technique can be mastered on a realistic model before actually administering to patients. The students can palpate the hand, which is made of supple, resilient plastic with a human feel. The vein is located, the needle introduced, and blood is withdrawn or fluid injected. The model can be used repeatedly. Complete with blood powder and carrying tray.
Product Family Options
Trainer
Informations sur le produit
  • Désignation
    Availability
    Price
  • IV Hand
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 286,40 $
    1 286,40 $
    Each
       1 286,40 $
    Supplier Number: 1005754
Replacement Materials
Informations sur le produit
  • Désignation
    Availability
    Price
  • Replacement Veins
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 90,75 $
    90,75 $
    Each
       90,75 $
    Supplier Number: 140-143
  • Replacement Reservoir Bag
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 47,90 $
    47,90 $
    Each
       47,90 $
    Supplier Number: 140-144