Sampling Tubes and Systems, Drager
À propos de cet article
Using the Drager sampling system, hazardous substances in the air are collected using a suitable medium via adsorption or chemisorption.
In active sampling, the air to be evaluated is drawn through a sampling tube such as Activated Charcoal and Silica Gel tube types with a pump and passive sampling is performed with diffusion samplers like the Dräger ORSA or the Dräger Nitrous Oxide diffusion samplers.
3 Options Available Below
Product Details & Documents
Using the Drager sampling system, hazardous substances in the air are collected using a suitable medium via adsorption or chemisorption.
In active sampling, the air to be evaluated is drawn through a sampling tube such as Activated Charcoal and Silica Gel tube types with a pump and passive sampling is performed with diffusion samplers like the Dräger ORSA or the Dräger Nitrous Oxide diffusion samplers.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 8101821
- Supplier Number: 6728631
- Supplier Number: 6728891
- Silica gel tubes, Type NIOSHDiscontinued-Supplier Number: -