Simulaids® Infant Intubation Trainer | Avantor
Simulaids® Infant Intubation Trainer
Supplier:   Nasco
Simulaids® Infant Intubation Trainer
Supplier:  Nasco
Certificates
Simulaids® Infant Intubation Trainer
Simulaids® Infant Intubation Trainer
2 Options Available Below
Product Details & Documents
Basic Infant Intubation Practice
  • Perform nasal or oral intubation
  • Includes carrying bag
This trainer of an infant allows practice and training of nasal and oral intubation. Features include: Inflatable lungs and stomach, Vocal cords are highlighted in white for easy laryngoscope viewing, Anatomical landmarks such as uvula, vocal cords, glottis, epiglottis, larynx, arytenoid cartilage, trachea, esophagus. Supplied with carrying bag and lubricant.
Product Family Options
Trainer
Informations sur le produit
  • Désignation
    Availability
    Price
  • Infant Intubation Head Trainer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,268.70
    $2,268.70
    Each
       $2,268.70
    Supplier Number: 101-115
Replacement Materials
Informations sur le produit
  • Désignation
    Availability
    Price
  • Replacement Lungs And Stomachs
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $117.15
    $117.15
    Each
       $117.15
    Supplier Number: 101-121