Condom Trainer
Condom Trainer
Condom Trainer
Condom Trainer
Condom Trainer
Demonstrate the proper use of condoms by using this realistic model.
  • Four stoma sites
  • Can be fitted with bags
  • Includes transport or storage case
Consists of an erect penis, 12 condoms, syringe and artificial semen (UV-fluorescent fluid) to simulate ejaculation. Mounted on a stand with suction cups and delivered with carrying bag.
Product Family Options
  • Désignation
  • Condom Trainer
    Each
    Each - $768.00
    $768.00
    Each
       $768.00
     : 1005560
  •  
     : -