Hand-Held I.U.D Trainer
I.U.D. Trainer
Catalog # 470119-230
Supplier: Nasco
Fournisseur #: 160-1912
Jump to:
Product Details & Documents
Economic Model Of Iud Placement
- Sized for easy storage
- Durable plastic construction
Hand-held trainer which is a suitable aid for understanding correct positioning of I.U.D. (Intrauterine Device) in the uterus. Made of durable plastic, the trainer features a transparent cover which allows easy visualization of insertion and placement of I.U.D. (I.U.D. not included).
Ordering information: I.U.D is not Included.
Spécifications
- Désignation:I.U.D. Trainer