Eisco® Root Tip Structure
Root Tip Structure, 1, 28 cm, 48 cm, 6 cm
Catalog # 470135-952
Supplier: Wards
Fournisseur #: BM0054
Jump to:
Product Details & Documents
Showing with a longitudinal section through tissue structure of wild rape root (snaps arvensis), 250 times magnified.
Spécifications
- Désignation:Root Tip Structure
- Hauteur:6 cm
- Longueur:48 cm
- Width:28 cm
- No. of Parts:1