Eisco® Color Coded Skull
Color Coded Skull, 3, 16 cm, 21 cm, 21 cm
470136-324
: Wards
: AMCH1009AS
Bright colors differentiate parts of the human skull.
- Calvarium cut
- 3 part skull
The skull cap is removable, and is held together with pegs.
- Désignation:Color Coded Skull
- Hauteur:21 cm
- Longueur:21 cm
- Width:16 cm
- No. of Parts:3