Eisco® Standing Urinary System
Standing Urinary System, 5, 23 cm, 33 cm, 36 cm
Catalog # 470136-316
Supplier: Wards
Fournisseur #: AM0356
Jump to:
Product Details & Documents
3D model of male urinary system and its blood supply is free standing, enabling study from all aspects.
1.5X life-size. The kidneys, adrenal and blood vessels, a section of the pelvic bone with pubic symphysis, lower portion of the bladder and prostate, the upper portion of the bladder and prostate.
Spécifications
- Désignation:Standing Urinary System
- Hauteur:36 cm
- Longueur:33 cm
- Width:23 cm
- No. of Parts:5