LevGo® smartSpatula® Disposable Lab Spatula
Catalogue # 470125-222
Fournisseur: Avantor
Spécifications
- Description:Disposable lab spatula
- Cat. no.:470125-222
- Supplier No.:WARD470125-222
Spécifications
À propos de cet article
Great for handling small amount of materials.
Utilitarian size: 210×7 mm (8.5×0.25"), 20/pack
- Spoon/scraper fits into 15 ml tube and scoop fits into 0.5 ml tube
-
100% polypropylene is autoclavable and resistant to dilute acids and bases
Blue contrasts with white material
Versatile double-ended polypropylene tool that can scoop, scrape, spoon, stir, collect, transfer, handle or manipulate a variety of chemicals and biological materials.
Ordering information:
The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc., patent 8,709,362.