Eisco® Numbered Human Skull | Avantor
Eisco® Numbered Human Skull
Numbered Human Skull, 3, 16 cm, 21 cm, 21 cm
Catalog # 470136-224
Supplier:   Wards
Fournisseur #:  AM0124AS
Eisco® Numbered Human Skull
Catalog # 470136-224
Supplier:  Wards
Eisco® Numbered Human Skull
Eisco® Numbered Human Skull
Numbered Human Skull, 3, 16 cm, 21 cm, 21 cm
Price
295,25 $
Log in pour la disponibilité
Each
Product Details & Documents
This model features numbered and removable parts to ease research.
  • Spring-Loaded Jaw
  • Removable Skullcap
  • Reference Sheet Included
The jaw is spring loaded, and the skullcap is removable for further interior inspection.
The model includes a sheet that identifies the name connected to the numbers on the parts.
Spécifications
  • Désignation:
    Numbered Human Skull
  • Hauteur:
    21 cm
  • Longueur:
    21 cm
  • Width:
    16 cm
  • No. of Parts:
    3