Life/form® Cath-Ed Training Models | Avantor
Life/form® Cath-Ed Training Models
Supplier:   Nasco
Life/form® Cath-Ed Training Models
Supplier:  Nasco
Certificates
Life/form® Cath-Ed Training Models
Life/form® Cath-Ed Training Models
2 Options Available Below
Product Details & Documents
Perfect Introduction For Catheter Training
  • Combines tactile and visual cues
  • Includes a functional bladder
  • Viewport to visualize proper foley catheter inflation
These unique Life/form® simulators combine visual and tactile learning sensations of catheterization in one compact unit. The mid-sagittal dissections allow detailed study of the internal anatomy and provide students an opportunity to follow what happens when they catheterize a patient. The removable plastic cover identifies key anatomical structures. With practice on Cath-Ed Simulators, normal restrictions at the bulbous urethra and bladder sphincter can be detected and identified. A Foley catheter can be demonstrated and seen through the viewing port. Includes a hard carrying case, lubricant, one catheter, 50 cc bulb, syringe, and instruction manual. These simulators do not have a functional bladder.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Cath-Ed Model Male
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,691.55
    $2,691.55
    Each
       $2,691.55
    Supplier Number: LF01025
  • Cath-Ed Model Female
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,691.55
    $2,691.55
    Each
       $2,691.55
    Supplier Number: LF01026