3B Scientific® MICROanatomy™ Bone Structure Model
Product Details & Documents
Detailed Bone Structure Along Multiple Planes.
- Three-Year Manufacturer's Warranty
- Includes 3B Smart Anatomy
The bone-structure model shows various planes in cross and longitudinal section through all levels of the bone as well as a two-plane section through the inner structure of the bone marrow. The model makes identifying the typical elements of a lamellar bone easy. It also helps students understand the bone's structure and function with the characteristic osteons, which are also referred to as Haversian systems.
This bone-structure model provides a graphic illustration of the interplay of individual components such as spongy and compact substance, endosteum, cortical substance, osteocytes, and Volkmann's and Haversian canals.
Informations sur les accessoires: For information about 3B Smart Anatomy, please see the document attached on this page.
Spécifications
- Désignation:Bone Structure
- No. of Parts:1