Life/form® Birthing Station Simulator | Avantor
Life/form® Birthing Station Simulator
Birthing Station Simulator
Catalog # 470119-244
Supplier:   Nasco
Fournisseur #:  LF01070
Life/form® Birthing Station Simulator
Catalog # 470119-244
Supplier:  Nasco
Life/form® Birthing Station Simulator
Life/form® Birthing Station Simulator
Birthing Station Simulator
Price
$3,964.85
Log in pour la disponibilité
Each
Product Details & Documents
Learn Palpation Techniques
  • Three head positions in relation to ischial spines
  • Includes carrying case
This birthing simulator is especially designed to demonstrate and practice the palpation of the birth canal. It shows the relationship between fetal head and ischial spines of the pelvic bone during birth. A simulated fetal head of the birth simulator can be locked in three different stations to demonstrate its position above, between and below the ischial spines. This anatomical birthing model is a great addition to any lesson about the human birth process. The birthing simulator is delivered with storage case. It is great for any gynecological and birth training.
Spécifications
  • Désignation:
    Birthing Station Simulator