Life/form® Central Venous Cannulation Simulator | Avantor
Life/form® Central Venous Cannulation Simulator
Supplier:   Nasco
Life/form® Central Venous Cannulation Simulator
Supplier:  Nasco
Certificates
Life/form® Central Venous Cannulation Simulator
Life/form® Central Venous Cannulation Simulator
1 Options Available Below
Product Details & Documents
Ideal Trainer Form Acls And Atls
  • Replicable muscle and bone layer
  • Landmarks exactly like a patient
  • Allows for use of a swan-ganz catheter
Designed for learning and practicing CVC techniques, the Life/form® Central Venous Cannulation Simulator is directed to the emergency medical field. ACLS and ATLS participants will find this simulator to be the ideal trainer. Anatomically accurate, palpation is exactly the same as on a patient. The sternal notch, sternocleidomastoid, clavicle, and other supporting features make this simulator a pleasure to work with. Internal features include a replaceable muscle and bone section, subclavian, internal jugular, external jugular, and carotid. Use of a Swan-Ganz catheter is possible and changing of the internal tubing is extremely easy. The neck of the simulator is positioned to the left, making landmark identity quite easy visually. The sternal notch “sinks” 1-1.5 cm when palpated. All external and internal landmarks correlate to a live patient. Modified to meet ATLS and ACLS standards, this simulator has proven itself in the emergency medical field. The consultant for this project was Forrest M. Gridley, M.A., P.A.-C. Includes fluid supply bags, replaceable skin, needles, one quart of Life/form® blood, and instruction guide in a hard carry case.
Ordering information: Fluid supply stand not included.
Product Family Options
Simulator
Informations sur le produit
  • Désignation
    Availability
    Price
  • Central Venous Cannulation Simulator
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,829.25
    $2,829.25
    Each
       $2,829.25
    Supplier Number: LF01087