Ranger® Industrial Bench Scale
: Ohaus
Top-of-the-line compact scales for even the most complex industrial applications. Simplify complex applications and minimize the need for manual calculations with ten advanced application modes, peripheral device control and an optional kit for a second scale platform.
- Very precise scale with one-second stabilization time, up to 75,000d or 350,000d display resolution and legal-for-trade certifications
- Extremely durable scale designed to thrive in rugged industrial environments
- Features IP54-rated metal housing and sealed metal terminal
For Ranger 7000, there is one resounding theme: it offers the best of every ingenious feature that make OHAUS industrial products the right tools for your toughest weighing jobs. Ranger 7000 boasts the highest resolution, largest display, most application modes and connectivity options, as well as the largest memory library of any industrial bench scale in the OHAUS portfolio.
Frequently Bought Together