DP Series Vacuum Drying Ovens, Yamato Scientific
Large-capacity vacuum drying oven with the possibility to form a system according to your purpose.
- Large scale treatment
- Decompressed chamber
- PID control by microprocessor
- Interactive key input device
- Graphic monitor
Vacuum drying oven for treatment on a large scale, designed for various purposes. Ovens feature decompressed chamber with direct heating and PID control by microprocessor (temperature control method). Equipped with an interactive key input device and graphic monitor. Various operational functions, including program operation and other add-on functions. A vacuum pump (not included) can be stored in the holder.
: Vacuum pump is required (not included). Includes 2 shelves.
- Catalog No:
- 76550-232
- 75846-554
- 75846-552
- Temperature control:
- ±1.0 °C (at 200 °C)
- ±1.0 °C (at 200 °C)
- ±1.0 °C (at 200 °C)
- Temperature range:
- 40…200 °C
- 40…200 °C
- 40…200 °C
Frequently Bought Together