Anti-GPATCH4 Rabbit Polyclonal Antibody
150 µL
10087-742
: 24399-1-AP-150UL
GPATCH4 (G patch domain-containing protein 4) is a 446 amino acid protein containing one G-patch domain. Existing as three alternatively spliced isoforms. This antibody is a rabbit polyclonal antibody raised against the 189 residues of N-terminal of human GPATCH4 protein
Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200
Type: Primary
Antigen: GPATCH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Taille:150 µL
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Antigen affinity Purification
- Antibody type:Primary
- Isotype:IgG
- Host:Rabbit
- Shipping temperature:Store at -20°C
- Antigen name:G Patch Domain Containing 4
- Cat. No.:10087-742
- Reactivity:Human|Rat|Mouse
- Antigen synonyms:GPATC4.|G patch domain containing protein 4|G patch domain containing 4
- Immunogen:Fusion Protein
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:GPATCH4
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- ELISA:Yes



