Order Entry
Canada
ContactUsLinkComponent
Anti-ACSM2A Rabbit Polyclonal Antibody
Catalogue # 10082-464
Fournisseur:  Proteintech
undefined
Anti-ACSM2A Rabbit Polyclonal Antibody
Catalogue # 10082-464
Fournisseur:  Proteintech
Supplier Number:  22862-1-AP-150UL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Acyl-CoA Synthetase Medium-chain Family Member 2A
  • Antigen symbol:
    ACSM2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    150 µL
  • Environmentally Preferable:
  • Antigen synonyms:
    A-923A4.1|ACSM2
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10082-464
  • Supplier No.:
    22862-1-AP

Spécifications

À propos de cet article

ACSM2A(Acyl-coenzyme A synthetase ACSM2A, mitochondrial) is also named as ACSM2, MACS2 and belongs to the ATP-dependent AMP-binding enzyme family. It acts as a medium-chain fatty acid : CoA ligase with the caculated molecular weight of 64 kDa and acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4-unsaturated acids.

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: ACSM2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human