Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Anti-AMPD1 Rabbit Polyclonal Antibody | Avantor

Anti-AMPD1 Rabbit Polyclonal Antibody

150 µL
Catalog # 10082-850
Supplier:   Proteintech
Fournisseur #:  19780-1-AP-150UL
Anti-AMPD1 Rabbit Polyclonal Antibody
Catalog # 10082-850
Supplier:  Proteintech
Anti-AMPD1 Rabbit Polyclonal Antibody
Anti-AMPD1 Rabbit Polyclonal Antibody

150 µL

Price
$717.22
Log in pour la disponibilité
Each (150µl)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

AMPD1, also named as MAD and MADA, belongs to the adenosine and AMP deaminases family. AMP deaminase plays a critical role in energy metabolism. It catalyzes the reaction: AMP + H2O = IMP + NH3. Defects in AMPD1 are the cause of adenosine monophosphate deaminase deficiency muscle type (AMPDDM) which is a metabolic disorder resulting in exercise-related myopathy. The antibody is specific to AMPD1.

Western Blot: Mouse Skeletal Muscle Tissue, 1:500-1:5000

Type: Primary
Antigen: AMPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Spécifications
  • Taille:
    150 µL
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Antigen affinity Purification
  • Antibody type:
    Primary
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Shipping temperature:
    Store at -20°C
  • Antigen name:
    AMP deaminase 1
  • Cat. No.:
    10082-850
  • Reactivity:
    Human|Rat|Mouse
  • Antigen synonyms:
    Ampd1|Myoadenylate deaminase|Ampd-1|AMP deaminase isoform M|AI553520
  • Immunogen:
    Peptide
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    AMPD1
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.