Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Anti-DUSP22 Rabbit Polyclonal Antibody | Avantor

Anti-DUSP22 Rabbit Polyclonal Antibody

150 µL
Catalog # 10081-734
Supplier:   Proteintech
Fournisseur #:  16514-1-AP-150UL
Anti-DUSP22 Rabbit Polyclonal Antibody
Catalog # 10081-734
Supplier:  Proteintech
Anti-DUSP22 Rabbit Polyclonal Antibody
Anti-DUSP22 Rabbit Polyclonal Antibody

150 µL

Price
$717.22
Log in pour la disponibilité
Each (150µl)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

DUSP22(Dual specificity protein phosphatase 22) is also named as JSP1, LMWDSP2, MKPX. It has been demonstrated to belong to a new subgroup of small DSPs anchored at the membranes by an N-terminal myristic acid moiety and DUSP22 play a regulatory role in the JNK or p38 MAPK pathways. It has 2 isoforms produced by alternative splicing with the molecular weight of 21 kDa and 23 kDa.

IHC: Human lung cancer Tissue, 1:20-1:200; Western Blot:Transfected HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: DUSP22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Spécifications
  • Taille:
    150 µL
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Antigen affinity Purification
  • Antibody type:
    Primary
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Shipping temperature:
    Store at -20°C
  • Antigen name:
    Dual Specificity Phosphatase 22
  • Cat. No.:
    10081-734
  • Reactivity:
    Human
  • Antigen synonyms:
    JSP1|LMWDSP2|MKPX|LMW-DSP2|JKAP|MKP-x|JSP-1|VHX
  • Immunogen:
    Recombinant Protein
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    DUSP22
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.