Anti-SYNCRIP Rabbit Polyclonal Antibody
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Synaptotagmin-binding, cytoplasmic RNA-interacting protein(SYNCRIP), also identified as hnRNP Q, involves in mRNA processing mechanisms. It's component of the CRD-mediated complex that promotes MYC mRNA stability. Also it is part of the APOB mRNA editosome complex and may modulate the postranscriptional C to U RNA-editing of the APOB mRNA through either by binding to A1CF (APOBEC1 complementation factor), to APOBEC1 or to RNA itself. May be involved in translationally coupled mRNA turnover. Implicated with other RNA-binding proteins in the cytoplasmic deadenylation/translational and decay interplay of the FOS mRNA mediated by the major coding-region determinant of instability (mCRD) domain. Interacts in vitro preferentially with poly(A) and poly(U) RNA sequences.
Western Blot:A431 Cells, 1:500-1:5000
Type: Primary
Antigen: SYNCRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Spécifications
- Taille:150 µL
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Antigen affinity Purification
- Antibody type:Primary
- Isotype:IgG
- Host:Rabbit
- Shipping temperature:Store at -20°C
- Antigen name:Synaptotagmin Binding, Cytoplasmic RNA Interacting Protein
- Cat. No.:10095-506
- Reactivity:Human|Rat|Mouse
- Immunogen:Recombinant Protein
- Conjugation:Unconjugated
- Antigen symbol:SYNCRIP
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



