Whatman™ Specialized Test Papers | Avantor
Whatman™ Specialized Test Papers
Supplier:   Cytiva
Whatman™ Specialized Test Papers
Supplier:  Cytiva
Certificates
Whatman™ Specialized Test Papers
Whatman™ Specialized Test Papers
3 Options Available Below
Product Details & Documents
Cytiva offers a range of pH indicator and test papers to meet specific needs.
  • Convenient and portable for field use
Potassium iodide test paper: For detecting chlorine and other oxidising agents. In acid solution, oxidising agents react with the iodide in the test paper to liberate iodine. The paper turns blue in the presence of an oxidizing agent (e.g., CI₂, Br₂, H₂O₂, HNO₂ etc.).
Lead acetate test paper: Used for detecting hydrogen sulfide, this rapid qualitative test paper when wetted with distilled water can detect as little as 5 ppm of H₂S in the atmosphere or in a gas stream. Hydrogen peroxide can be detected with this paper by preblackening the paper in H₂S. Concentrations as low as 4 ppm can be detected.
These high-quality products combine ease-of-use with unsurpassed accuracy and consistency.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Lead acetate test paper, 7 mm×50 m, with dispenser
    1 Reel
    Discontinued
    Supplier Number: 2602-501A
  • Potassium iodide test paper, reel 7 mm×5 m, with dispenser
    1 Reel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $49.11
    $49.11
    Pack of 1
       $49.11
    Supplier Number: 2602-500A
  • Potassium iodide test paper, reel 7 mm×5 m, with dispenser
    1 Reel
    Discontinued
    Supplier Number: 2602-500A
  • Lead acetate test paper, 7 mm×50 m, with dispenser
    1 Reel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $54.97
    $54.97
    Pack of 1
       $54.97
    Supplier Number: 2602-501A
  • Starch iodide test papers, 10 books of 20 strips
    200 strips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $314.11
    $314.11
    Pack of 100
       $314.11
    Supplier Number: 2651-500
  • Starch iodide test papers, 10 books of 20 strips
    200 strips
    Discontinued
    Supplier Number: 2651-500