Wescorp ESD High Temperature Polymide Tape, Desco
Fournisseur: Desco
Certificats
À propos de cet article
Used in applications masking PCB gold features for wave soldering and IR reflow ovens
- Removal leaves little or no residue
- Static charge generation during unwind and peel off stainless steel plate application is 5 volts @ 50% RH
- 1.0 mil thick (0.001" or 0.0254 mm) Polyimide Film (DuPont Kapton® or equivalent)
- 1.4 mil thick (0.0014" or 0.0356 mm) conductive polysiloxide adhesive
- Adhesive surface resistivity 10E3 to 10E4 ohms
- Max Temperature 260°C (500°F)
- For best results, apply to board using a rubber roller
Tapes should be stored in a dry, well ventilated room with a reasonably consistent temperature of 68°F (20°C) and be protected from exposure to direct sunlight.
Frequently Bought Together