Optical Dissolved Oxygen Meter, smarTROLL RDO, In-Situ
Fournisseur: IN-SITU, INC.
Certificats
À propos de cet article
The smarTROLL Rugged Dissolved Oxygen (RDO) Handheld combines optical dissolved oxygen (DO) technology with revolutionary smartphone mobility
Simultaneously measure four parameters: Dissolved oxygen with the optical RDO Sensor, Barometric pressure, Air temperature and Water temperature. Get instant results—No warm-up or stabilization time required. Calibrations last for months.
Waterproof case is availabe for your iPhone and iPod touch device.
The intuitive iSitu® application eliminates training time and runs on your iPhone®, iPod touch®, or iPad® device.
Certifications: FCC and EPA Approved. CE compliant.
Ordering information: Bundles include probe (RDO, temperature sensor), weighted guard, battery pack, specified cable length, calibration cup and Operator’s Manuals on CD. Does not include iOS device.
Frequently Bought Together