VWR® pH/Temperature Meter, Pen Style
Catalog # 74730-030
Supplier: VWR International
Specifications
- Temperature range:0.0 to 99.9°C (32.0 to 211.8°F)
- Description:pH10A Meter
- Temperature accuracy:±0.3 (±0.6°F)
- Temperature resolution:0.1 °C (0.2 °F) °C
- Display:98 × 64 graphic LCD
- Weight:105 g (3.7 oz.)
- pH accuracy:±0.02 pH ± 1 LSD, ±0.1 (within 10°C of calibration), ±0.2 (within 20°C of calibration)
- pH range:0.00 to 14.00
- pH resolution:0.01
- Cat. no.:74730-030
Specifications
About this item
The model pH10A meter is the perfect instrument for economical spot sampling of pH and temperature in applications such as wastewater, surface water, aquaculture, hydroponics, pools, and education.
- Automatic calibration and buffer recognition
- 50-set memory (pH, temp, date and time stamp)
- Replaceable single- or double-junction electrode
- GLP functionality
- 1-, 2-, or 3-point calibration
- Hold feature locks stable readings on the display
- >200 hour battery life with low battery indicator
This ultra-compact instrument even includes a graphic display with on-screen instructions. Available with single- or dual-junction electrodes (comes with single-junction electrode). No need to throw this pen-style pH instrument away when the sensor goes bad like some other products. Simply replace the electrode within minutes and keep testing.
Certifications: Waterproof IP-67. CE compliance.
Ordering information: Instrument includes model 606110 pH electrode, batteries, and instruction manual. Manufacturer's one-year warranty.