Order Entry
Canada
ContactUsLinkComponent
Anti-IgM µ Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalogue # 10152-042
Fournisseur:  Immunoreagents
undefined
Anti-IgM µ Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalogue # 10152-042
Fournisseur:  Immunoreagents
Supplier Number:  GTXHU-006-DHRPX
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin M mu-chain
  • Antigen symbol:
    IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    1.0 mg
  • Epitope:
    µ-chain
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Lyophilized
  • Storage buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
  • Storage temperature:
    2-8 °C
  • Concentration:
    1.0 mg/ml (E 1% at 280 nm = 13.0)
  • Shipping temperature:
    2-8 °C
  • Immunogen:
    Human IgM, µ chain
  • Purification:
    Affinity purified using solid phase Human IgM
  • Cat. no.:
    10152-042
  • Supplier No.:
    IR1818

Spécifications

À propos de cet article

ImmunoReagents’ Horseradish Peroxidase (HRP) antibody conjugates are the most popular conjugates due to their high level of sensitivity, stability, and low costs. HRP conjugates are an ideal choice for use in ELISA, Immunohistochemistry (IHC), and Western blotting experiments.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Human