Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody | Avantor

Anti-IgG Goat Polyclonal Antibody

1.0 mg
Catalog # 10149-774
Supplier:   Immunoreagents
Fournisseur #:  GTXHU-004-F2
Anti-IgG Goat Polyclonal Antibody
Catalog # 10149-774
Supplier:  Immunoreagents
Anti-IgG Goat Polyclonal Antibody
Anti-IgG Goat Polyclonal Antibody

1.0 mg

Price
$204.52
Log in pour la disponibilité
Each (1mg)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Human

Spécifications
  • Taille:
    1.0 mg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Flow cytometry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Epitope:
    F(c)
  • Form:
    Clear, colorless liquid, 0.2 µm filtered
  • ImmunoFluorescence:
    Yes
  • Storage temperature:
    2-8 °C
  • Antigen name:
    Immunoglobulin G
  • Antibody fragment:
    F(c)
  • Format:
    Purified IgG
  • Immunogen:
    Human IgG, Fc fragment
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Storage buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • Purification:
    Affinity purified using solid phaseHuman IgG
  • Antibody type:
    Secondary
  • Isotype:
    IgG
  • Host:
    Goat
  • Concentration:
    > 4.5 mg/ml (E 1% at 280 nm = 13.0)
  • Cross adsorption:
    Yes
  • Shipping temperature:
    2-8 °C
  • Cat. No.:
    10149-774
  • Reactivity:
    Human
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    IgG F(c)
  • ELISA:
    Yes
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.