Mouse Affinity Purification Resin, ImmunoReagents
Mouse Affinity Purification Resin, ImmunoReagents
Purified Mouse IgG Coupled to 4% Agarose Beads, 2 mL
Catalog # 10148-980
Supplier:   Immunoreagents
Supplier #:  GE-017
Mouse Affinity Purification Resin, ImmunoReagents
Catalog # 10148-980
Supplier:  Immunoreagents
CAS Number:  
Mouse Affinity Purification Resin, ImmunoReagents
Mouse Affinity Purification Resin, ImmunoReagents
Purified Mouse IgG Coupled to 4% Agarose Beads, 2 mL
CAS Number:  
Price
$714.41
Log in pour la disponibilité
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
ImmunoReagents Mouse IgG Affinity Prep resins are prepared using highly purified species specific immunoglobulins
Immunoglobulins demonstrate a single arc when analyzed against normal serum proteins by IEP. The resins provided comprise of immunoglobulins covalently bound to a 4% agarose bead utilizing strong and stable hydrazide/aldehyde group chemistry. The Affinity Prep resins have 8-10 mgs of antibodies per/ml of resin, with a 24 month shelf life reagents.
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulin s from various species cross absorbed to provide high specificity.  
This resin is suitable for removal of proteins recognizing Mouse IgG from liquid solutions, including traditional Immunoabsorptions and Immunoprecipitation protocols.
Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: ImmunoReagents products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles. Additionally, products can by customized based on specific assay requirements. Contact your Avantor representative for more information.
Spécifications
Spécifications
  • Désignation:
    Purified Mouse IgG Coupled to 4% Agarose Beads
  • Taille:
    2 mL
  • Cat. No.:
    10148-980
  • Storage temperature:
    2 to 8°C, Do Not Freeze