Glas-Col® Heating Tapes, Ribbon-Type Elements
À propos de cet article
GLAS-COL® Tape heats odd lengths of glass or metal pipe
Ribbon-type heating elements running lengthwise in the tape are encased in durable glass braid and are impregnated and covered with tough silicone rubber.
The silicone rubber insulates stranded terminal leads at one end and waterproofs the fittings at both ends. It also permits use in high-moisture areas and helps to make the tapes resistant to most laboratory environment conditions. All tapes are supplied with a 2 blade plug and have a temperature limit of 249 °C (480 °F).
: Note: Power ratings listed are for tapes not covered by insulation when installed. If covered, temperature should be monitored and controlled to avoid exceeding the limit. Short narrow tapes that are rated at less than 115 volt must be operated with a variable voltage transformer.
14 Options Available Below
Product Details & Documents
GLAS-COL® Tape heats odd lengths of glass or metal pipe
Ribbon-type heating elements running lengthwise in the tape are encased in durable glass braid and are impregnated and covered with tough silicone rubber.
The silicone rubber insulates stranded terminal leads at one end and waterproofs the fittings at both ends. It also permits use in high-moisture areas and helps to make the tapes resistant to most laboratory environment conditions. All tapes are supplied with a 2 blade plug and have a temperature limit of 249 °C (480 °F).
: Note: Power ratings listed are for tapes not covered by insulation when installed. If covered, temperature should be monitored and controlled to avoid exceeding the limit. Short narrow tapes that are rated at less than 115 volt must be operated with a variable voltage transformer.
Product Family Options
Informations sur le produit
- DésignationLongueurLargeurElectricalAvailabilityPrice
- Heating Tapes, Ribbon-Type Elements122 cm (4 ft.)6.4 mm (¼")45 V, 60 WLog in pour la disponibilitéEach$387.13Supplier Number: CG-14050-01
- Heating Tapes, Ribbon-Type Elements182.8 cm (6 ft.)6.4 mm (¼")70 V, 90 WLog in pour la disponibilitéEach$421.57Supplier Number: CG-14050-02
- Heating Tapes, Ribbon-Type Elements304.80 cm (10 ft.)6.4 mm (¼")120 V, 150 WLog in pour la disponibilitéEach$507.00Supplier Number: CG-14050-04
- Heating Tapes, Ribbon-Type Elements122 cm (4 ft.)12.7 mm (½")45 V, 140 WLog in pour la disponibilitéEach$406.05Supplier Number: CG-14050-05
- Heating Tapes, Ribbon-Type Elements182.8 cm (6 ft.)12.7 mm (½")70 V, 210 WLog in pour la disponibilitéEach$466.44Supplier Number: CG-14050-06
- Heating Tapes, Ribbon-Type Elements243.8 cm (8 ft.)12.7 mm (½")95 V, 280 WLog in pour la disponibilitéEach$526.65Supplier Number: CG-14050-07
- Heating Tapes, Ribbon-Type Elements304.8 cm (10 ft.)12.7 mm (½")115 V, 340 WLog in pour la disponibilitéEach$615.00Supplier Number: CG-14050-08
- Heating Tapes, Ribbon-Type Elements152.4 cm (5 ft.)15.9 mm (⅝")115 V, 145 WLog in pour la disponibilitéEach$542.99Supplier Number: CG-14050-10
- Heating Tapes, Ribbon-Type Elements182.8 cm (6 ft.)15.9 mm (⅝")115 V, 120 WLog in pour la disponibilitéEach$571.09Supplier Number: CG-14050-11
- Heating Tapes, Ribbon-Type Elements213.4 cm (7 ft.)15.9 mm (⅝")115 V, 105 WLog in pour la disponibilitéEach$623.00Supplier Number: CG-14050-12
- Heating Tapes, Ribbon-Type Elements91.4 cm (3 ft.)25.4 mm (1")115 V, 150 WLog in pour la disponibilitéEach$548.71Supplier Number: CG-14050-16
- Heating Tapes, Ribbon-Type Elements122 cm (4 ft.)25.4 mm (1")115 V, 120 WLog in pour la disponibilitéEach$580.45Supplier Number: CG-14050-17
- Heating Tapes, Ribbon-Type Elements152.4 cm (5 ft.)25.4 mm (1")115 V, 100 WLog in pour la disponibilitéEach$654.24Supplier Number: CG-14050-18
- Heating Tapes, Ribbon-Type Elements304.8 cm (10 ft.)25.4 mm (1")115 V, 700 WLog in pour la disponibilitéEach$853.00Supplier Number: CG-14050-19