ContactUsLinkComponent
Guinea Pig Serum, ImmunoReagents
Guinea Pig Serum, ImmunoReagents
Supplier:  Immunoreagents
Certificates
undefined
Guinea Pig Serum, ImmunoReagents
Supplier:  Immunoreagents
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Catalog No:
  • 10152-216
  • 10152-214
  • Application:
  • ELISA, WB, IHC, IP FACS, ID and Labeling
  • ELISA, WB, IHC, IP FACS, ID and Labeling
  • Storage temperature:
  • Store lyophilized material at 2-8 °C. For long term storage after reconstitution, prepare small aliquots and store at -20 °C . For storage at 2-8 °C, add a preservative to prevent growth of bacteria.
  • Store lyophilized material at 2-8 °C. For long term storage after reconstitution, prepare small aliquots and store at -20 °C . For storage at 2-8 °C, add a preservative to prevent growth of bacteria.