À propos de cet article
Shippers have been specifically designed by life science customers, for life science customers, and are the perfect solution for the transport of your high-value, temperature-sensitive, pharmaceutical, biological, and vaccine products.
- Ideal for transporting high-value items
- EasyLoc™ design
- Minimal packing components
- Maintains consistent temperature
- Three temperature ranges available
- 5-Day duration
Featuring an all new exclusive EasyLoc™ design, shippers have several unique characteristics, including self-supporting, light-weight insulated panels; minimal packing components; simple and convenient one-person assembly system; superior payload capacity of at least 1.0 cubic meter; optimized exterior dimensions ideal for airline transport; durable and reusable panels with interlocking edges, locking out air penetration; and ribbed design on panels to enhance convective airflow, helping to maintain a consistent temperature throughout.
Maximize your air freight utilization with Sonoco ThermoSafe’s LD7 Quarter PMC. The newest pallet solution to our fleet includes the groundbreaking ConvecTECH™ technology and unique “L” shaped corner panels to simplify assembly and enhance thermal performance. Available in three temperature ranges: 2 to 8°C Refrigerated, 15 to 25°C CRT, less than -20°C Frozen.