Granular dessicant, Sicapent® with indicator, MilliporeSigma
Supplier: MilliporeSigma
Certificates
About this item
Sicapent is a granular dessicant comprising ~75% phoshorus pentoxide on an inert inorganic carrier.
- Ease of use
- Fast water adsorption due to large surface area
- Colour indicator - colourless to green to blue
- Remains in particle form after 100% water uptake
1.00543.2800 replaces and is the same as SX0137-1. Part # Change only, Same Pack Size, Same Specifications.
Appliations include drying liquids, filling drying tubes, particularly for fast flowing gases in drying tubes
Bulk density: ~300 g/l
Frequently Bought Together