Absorbent, Chemizorb® Granules, EMD Millipore
Catalogue # CA1.01568.5000
Fournisseur: MilliporeSigma
Spécifications
- Description:Chemizorb® Granules
- Size:5 kg
- Cat. no.:CA1.01568.5000
- Packaging:Plastic bucket
Spécifications
À propos de cet article
Chemizorb® is a coarse grained material based on chemically inert porous mineral or synthetic copolymers (grain size >1 mm (95%), bulk density approx. 400 kg/m³). It is insoluble in water and in all other media that are liquid at room temperature. Both Chemizorb® granules and powder are suited for absorbing alkalis, acids and also heating bath liquids.
- Able to absorb up to 100% of its own weight
- Low viscosity liquids are absorbed instantaneously, highly viscous media (e.g. glycerine, oils) within a few minutes
- Good dosing is possible thanks to the coarse grained format
- Granule form is used in all those places where it is not suitable to use the powder form, for example in draughty rooms or outside
Merck products are not available from Avantor in all countries. Please check with your local distributor.