Whatman™ Extraction Thimbles for DIONEX ASE, Whatman products (Cytiva)
: Cytiva
For DIONEX ASE 200 extraction cell
- Ideal for all automated extraction apparatus
- Maximum temperature: 120 °C
Cellulose material. Grade 603.
- Catalog No:
- CA11013-406
- CA11013-268
- Max. operating temperature:
- 130 °C
- 130 °C
- Wall thickness:
- 1 mm
- 1 mm
Frequently Bought Together