Labx® 123 Low-Linting Wipers, Berkshire
Fournisseur: Berkshire
Certificats
À propos de cet article
Labx 123 is engineered as a lightweight low lint wiper.
It is 100% biodegradable and made from 100% manila hemp. Labx 123 is an environmentally friendly wipe that has been designed for ISO Class 6 (Class 1000) and above cleanroom applications.
Frequently Bought Together