Whatman™ Polyester Drain Discs
Whatman™ Polyester Drain Discs
Supplier:   Cytiva
Whatman™ Polyester Drain Discs
Supplier:  Cytiva
Certificates
Whatman™ Polyester Drain Discs
Whatman™ Polyester Drain Discs
5 Options Available Below
Product Details & Documents
The polyester drain disc is binder free and has a thickness of 100 µm.
  • Flat surface to eliminate filter tearing or rupturing
  • Used as a separator between membrane layers in serial stack filtration applications
This chemically inert support disc is available in a variety of diameters for use in a range of devices. With Application in General laboratory microfiltration, Quality control and sterility testing, Removal of particulates from HPLC solvents, Tissue culture media filtration.
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Épaisseur
    Availability
    Price
  • Polyester drain disc, PE, membrane filter accessories
    10 mm
    100 µm
    Discontinued
    Supplier Number: 230300
  • Polyester drain disc, PE, membrane filter accessories
    10 mm
    100 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $110.60
    $110.60
    Pack of 100
       $110.60
    Supplier Number: 230300
  • Polyester drain disc, PE, membrane filter accessories
    22 mm
    100 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $202.65
    $202.65
    Pack of 100
       $202.65
    Supplier Number: 230500
  • Polyester drain disc, PE, membrane filter accessories
    22 mm
    100 µm
    Discontinued
    Supplier Number: 230500
  • Polyester drain disc, PE, membrane filter accessories
    25 mm
    100 µm
    Discontinued
    Supplier Number: 230600
  • Polyester drain disc, PE, membrane filter accessories
    25 mm
    100 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $122.37
    $122.37
    Pack of 100
       $122.37
    Supplier Number: 230600
  • Polyester drain disc, PE, membrane filter accessories
    37 mm
    100 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $222.53
    $222.53
    Pack of 100
       $222.53
    Supplier Number: 230800
  • Polyester drain disc, PE, membrane filter accessories
    37 mm
    100 µm
    Discontinued
    Supplier Number: 230800
  • Polyester drain disc, PE, membrane filter accessories
    47 mm
    100 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $258.81
    $258.81
    Pack of 100
       $258.81
    Supplier Number: 231100
  • Polyester drain disc, PE, membrane filter accessories
    47 mm
    100 µm
    Discontinued
    Supplier Number: 231100