Eisco® NextGen Bunsen Burner with Flame Stabilizer
Eisco® NextGen Bunsen Burner with Flame Stabilizer
Supplier:   Wards
Eisco® NextGen Bunsen Burner with Flame Stabilizer
Supplier:  Wards
Certificates
Eisco® NextGen Bunsen Burner with Flame Stabilizer
Eisco® NextGen Bunsen Burner with Flame Stabilizer
2 Options Available Below
Product Details & Documents
The next generation in laboratory burners. NextGen Bunsen Burners are designed to be the safest on the market.
  • Designed for safety
  • Expanded cast iron base prevents easy tipping, even with hoses connected
  • Spaced feet so burner fits comfortably under stands
  • Candle stick style handle allows user to move burner without getting harmed
  • Features air flow adjustment sleeve and brass flame stabilizer
  • Base is coated with a chemical resistant powder coating
This basic Bunsen burner produces a safe continuous stream of flammable gas used for heating, sterilization and combustion. The burner has one adjustment which is an air flow adjustment sleeve; the flame is controlled via the valve at the gas output, not at the burner. A brass flame stabilizer is also included. This is a very basic, easy to use burner. Ribbed fuel input tube is 8 mm outside diameter, and versions jetted for liquid propane and natural gas are both available. Liquid propane burners have a BTU range of approx. 2000 to 3200. Maximum Temperature is approximately 1550 °C (2822° F). Natural gas burners have a BTU range of approx. 800 to 1200. Maximum temperature is approximately 1500 °C (2732° F).
Ordering information: Neoprene connecting hoses, 60 or 90 cm (items 470121-742 and 470220-806), recommended.
Product Family Options
Product Information
  • Désignation
    For
    Tube Diameter
    Hauteur
    Weight
    Availability
    Price
  • Burner with flame stabilizer
    Natural gas
    13 mm
    160 mm
    290 g
    Log in for availability
    Log in for availability
    Each
    Each - $58.00
    $58.00
    Each
       $58.00
    Supplier Number: CH0992NG
  • Burner with flame stabilizer
    Liquid petroleum
    13 mm
    160 mm
    290 g
    Log in for availability
    Log in for availability
    Each
    Each - $58.00
    $58.00
    Each
       $58.00
    Supplier Number: CH0992LP
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Neoprene connecting hose, 60 cm
    Neoprene connecting hose, 60 cm
    470121-742
    Log in for availability
    Each - $33.65