Eisco® NextGen Basic Bunsen Burner | Avantor
Eisco® NextGen Basic Bunsen Burner
Supplier:   Wards
Eisco® NextGen Basic Bunsen Burner
Supplier:  Wards
Certificates
Eisco® NextGen Basic Bunsen Burner
Eisco® NextGen Basic Bunsen Burner
2 Options Available Below
Product Details & Documents
The next generation in laboratory burners. NextGen Bunsen Burners are designed to be the safest on the market.
  • Designed for safety
  • Expanded base prevents easy tipping, even with hoses connected
  • Spaced feet so burner fits comfortably under stands
  • Candle stick style handle allows user to move burner without getting harmed
  • Features air flow adjustment sleeve
  • Base is coated with a chemical resistant powder coating
This basic Bunsen burner is used for safe continuous stream of flammable gas used for heating, sterilization and combustion. This burner has one adjustment which is an air flow adjustment sleeve; the flame is controlled via the valve at the gas output, not at the burner. This is a very basic, easy to use burner. Ribbed fuel input tube is 8 mm outside diameter, and versions jetted for liquid propane and natural gas are both available. Liquid propane burners have a BTU range of approx. 2000 to 3200. Maximum Temperature is approximately 1550 °C (2822 °F). Natural gas burners have a BTU range of approx. 800 to 1200. Maximum temperature is approximately 1500 °C (2732 °F).
Ordering information: Neoprene connecting hoses, 60 or 90 cm (items 470121-742 and 470220-806), recommended.
Product Family Options
Informations sur le produit
  • Désignation
    For
    Tube Diameter
    Hauteur
    Weight
    Availability
    Price
  • Basic Bunser burner
    Liquid petroleum
    11 mm
    145 mm
    270 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $45.90
    $45.90
    Each
       $45.90
    Supplier Number: CH0991LP
  • Basic Bunser burner
    Natural gas
    11 mm
    145 mm
    270 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $45.90
    $45.90
    Each
       $45.90
    Supplier Number: CH0991NG
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Neoprene connecting hose, 60 cm
    Neoprene connecting hose, 60 cm
    470121-742
    Log in pour la disponibilité
    Each - $33.65