KIMBLE® Thermometer Inlet Adapter with Hose Connection, DWK Life Sciences
Catalogue # KT197000-2440
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- ST Joint:24/40
- Environmentally Preferable:
- Cat. no.:KT197000-2440
- Overall Height:155 mm
- Fits Tubing I.D.:⅜"
- Supplier No.:197000-2440
Spécifications
À propos de cet article
Adapter features a hose connection on the side.
- Side hose connection
- Accommodates thermometer
- Manufactured from borosilicate glass
The hose connection on the side accepts 3/8” ID flexible tubing.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.