Transfer Buffer, Azure Biosystems
Catalogue # 10147-348
Fournisseur: Azure Biosystems
Product available on GSA Advantage®
Numéro CAS:
Spécifications
- Description:50X Concentrated Solution, Sufficient for 50 - 100 Blots
- Cat. no.:10147-348
- Supplier No.:AC2127
Spécifications
À propos de cet article
Azure’s proprietary transfer buffer provides improved detection of low-abundance and post-translationally modified proteins.
Transfer times can be reduced with this buffer relative to traditional transfer buffers, saving you time.
High-efficiency protein transfer and increased protein retention on the membrane add up to more sensitive Western blots.